Human recombinant profilin-2 (from E. coli)
Fournisseur: ProSci Inc.
92-202
PRSI92-202EA 306 EUR InStock
PRSI92-202
Human recombinant profilin-2 (from E. coli)
Protéines et Peptides
Profilin-II (PFN2) is ubiquitous protein which belongs to the profilin family. PFN2 binds to actin, thenaffects the structure of the cytoskeleton. At high concentrations, profiling prevents the polymerisation of actin, while increases that at low concentrations. PFN2 is a ubiquitous actin monomer-binding protein. It regulates actin polymerisation in response to extra cellular signals. PFN2 binds to PIP2; it inhibits the formation of IP3 and DG.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité