Human recombinant UCMA (from E. coli)
Fournisseur: ProSci Inc.
92-203
PRSI92-203EA 789 EUR InStock
PRSI92-203
Human recombinant UCMA (from E. coli)
Protéines et Peptides
C10orf49 is a secreted protein which encoded by the UCMA gene. It is a member of the UCMA family. C10orf49 is predominantly expressed in resting chondrocytes. It may be involved in the negative control of osteogenic differentiation of osteochondrogenic precursor cells in peripheral zones of fetal cartilage and at the cartilage-bone interface.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité