Human recombinant Arginase 1 (from E. coli)
Fournisseur: ProSci Inc.
92-205
PRSI92-205EA 761 EUR InStock
PRSI92-205
Human recombinant Arginase 1 (from E. coli)
Enzymes
ARG1 is a member of the ureohydrolase family of enzymes. ARG1 can catalyse the hydrolysis of arginine to ornithine and urea. In the urea cycle, ARG1 catalyses the fifth and final step, a series of biochemical reactions in mammals during which the body disposes of harmful ammonia. ARG1 is a cytosolic enzyme and expressed widely in the liver as part of the urea cycle. Inherited deficiency of this ARG1 causes argininemia, which is an autosomal recessive disorder characterised by hyperammonemia.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité