Human recombinant Ornithine decarboxylase (from E. coli)
Fournisseur: ProSci Inc.
92-208
PRSI92-208EA 584 EUR InStock
PRSI92-208
Human recombinant Ornithine decarboxylase (from E. coli)
Enzymes
Ornithine decarboxylase (ODC1) is an enzyme which belongs to the Orn/Lys/Arg decarboxylase class-II family. ODC1 is the rate-limiting enzyme of the polyamine biosynthesis pathway which catalyses ornithine to putrescine. The activity level for the enzyme varies in response to growth-promoting stimuli and exhibits a high turnover rate in comparison to other mammalian proteins.
Fusion-Tag: C-6 His, N-T7 tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité