Human recombinant sterol O-acyltransferase 2 (from E. coli)
Fournisseur: ProSci Inc.
92-209
PRSI92-209EA 789 EUR InStock
PRSI92-209
Human recombinant sterol O-acyltransferase 2 (from E. coli)
Enzymes
ACAT2 is a cytoplasmic enzyme which belongs to the thiolase family. ACAT2 takes part in lipid metabolism, lipoprotein assembly, catalyzing cholesterol esterification in mammalian cells. It is responsible for the synthesis of cholesteryl esters which are part of lipoproteins containing apoB. ACAT2 deficiency contributes to severe mental retardation and hypotonus.
Fusion-Tag: N-Trx, 6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité