Human Recombinant NHP2 (from E. coli)
Fournisseur: ProSci Inc.
92-211
PRSI92-211EA 654 EUR InStock
PRSI92-211
Human Recombinant NHP2 (from E. coli)
Protéines et Peptides
NHP2 belongs to the H/ACA snoRNPs gene family. snoRNPs are involved in various aspects of rRNA processing and modification and have been organised into 2 families: C/D and H/ACA. NHP2 forms a small ribonucleoprotein particle with GAR1 (NOLA1). NHP2 is involved in a variety of aspects of rRNA processing and modification. NHP2 localises to the dense fibrillar component of the nucleolus and in nuclear Cajal bodies. NHP2 may also be required for correct processing or intranuclear trafficking of TERC, the RNA component of the telomerase reverse transcriptase (TERT) holoenzyme.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité