Human recombinant Copine-1 (from E. coli)
Fournisseur: ProSci Inc.
92-213
PRSI92-213EA 620 EUR InStock
PRSI92-213
Human recombinant Copine-1 (from E. coli)
Protéines et Peptides
Copine-1(CPNE1) encodes a calcium-dependent protein which belongs to the copine family. CPNE1contains two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. However, CPNE1 does not contain a predicted signal sequence or transmembrane domains. CPNE1 may regulate molecular events at the interface of the cell membrane and cytoplasm. CPNE1 has a broad tissue distribution and it may function in membrane trafficking.
Fusion-Tag: N, C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité