Human recombinant Peptidase D (from E. coli)
Fournisseur: ProSci Inc.
92-214
PRSI92-214EA 789 EUR InStock
PRSI92-214
Human recombinant Peptidase D (from E. coli)
Enzymes
PEPD belongs to the peptidase M24B family of Eukaryotic-type prolidase subfamily. PEPD is a cytosolic dipeptidase that hydrolyses dipeptides with proline or hydroxyproline at the carboxy terminus. It is important in collagen metabolism because of the high levels of imino acids. Defects in PEPD are a cause of prolidase deficiency which is an autosomal recessive disorder associated with iminodipeptiduria.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité