Human recombinant TNF alpha (from E. coli)
Fournisseur: ProSci Inc.
92-217
PRSI92-217EA 852 EUR InStock
PRSI92-217
Human recombinant TNF alpha (from E. coli)
Protéines et Peptides
tumour Necrosis Factor- alpha (TNF- alpha) is secreted by macrophages, monocytes, neutrophils, T-cells, and NK-cells following stimulation by bacterial LPS. Cells expressing CD4 secrete TNF- alpha while cells that express CD8 secrete little or no TNF- alpha. Synthesis of TNF- alpha can be induced by many different stimuli including interferons, IL2, and GM-CSF. The clinical use of the potent anti-tumour activity of TNF- alpha has been limited by the proinflammatory side effects such as fever, dose-limiting hypotension, hepatotoxicity, intravascular thrombosis, and hemorrhage. Designing clinically applicable TNF- alpha mutants with low systemic toxicity has been of intense pharmacological interest. Human TNF- alpha that binds to murine TNF-R55 but not murine TNF-R7, exhibits retained anti-tumour activity and reduced systemic toxicity in mice compared with murine TNF- alpha, which binds to both murine TNF receptors. Based on these results, many TNF- alpha mutants that selectively bind to TNF-R55 have been designed. These mutants displayed cytotoxic activities on tumour cell lines in vitro and have exhibited lower systemic toxicity in vivo. Recombinant Human TNF- alpha High Active Mutant differs from the wild-type by amino acid subsitution of amino acids 1-7 with Arg8, Lys9, Arg10 and Phe157. This mutant form has been shown to have increased activity with less inflammatory side effects in vivo.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité