Human recombinant IL1 beta (from E. coli)
Fournisseur: ProSci Inc.
92-219
PRSI92-219EA 654 EUR InStock
PRSI92-219
Human recombinant IL1 beta (from E. coli)
Protéines et Peptides
IL1B belongs to the IL-1 family. Interleukin 1 (IL-1) is a family of polypeptide cytokines consisting of two agonists, IL-1 alpha (IL-1F1) and IL-1 beta (IL-1F2) encoded by two distinct genes and perform identical biological functions. IL-1 stimulates thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, and fibroblast growth factor activity. IL-1 proteins are involved in the inflammatory response. It is identified as endogenous pyrogens, and is reported to stimulate the release of prostaglandin and collagenase from synovial cells.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité