Human recombinant CaM kinase ID (from E. coli)
Fournisseur: ProSci Inc.
92-220
PRSI92-220EA 789 EUR InStock
PRSI92-220
Human recombinant CaM kinase ID (from E. coli)
Enzymes
CAMK1D is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. It contains a protein kinase domain. CAMK1D expressed in polymorphonuclear leukocytes and may be part of the chemokine signal transduction pathway that regulates granulocyte function. It may also be involved in modulation of neuronal apoptosis. It activates CREB-dependent gene transcription, regulates calcium-mediated granulocyte function and respiratory burst and promotes basal dendritic growth of hippocampal neurons.
Fusion-Tag: N-GST tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité