Human recombinant EIF5A2 (from E. coli)
Fournisseur: ProSci Inc.
92-221
PRSI92-221EA 620 EUR InStock
PRSI92-221
Human recombinant EIF5A2 (from E. coli)
Protéines et Peptides
EIF5A2 is a member of the eIF-5A family. It is a 153 amino acids protein that in humans is encoded by the EIF5A2 gene. EIF5A2 is expressed in ovarian and colorectal cancer cell lines and highly expressed in testis.It has an important function at the level of mRNA turnover and mediates effects of polyamines on neuronal process extension and survival. It plays an important role in brain development and function, and in skeletal muscle stem cell differentiation.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité