Human recombinant 4-1BB Ligand (from E. coli)
Fournisseur: ProSci Inc.
92-225
PRSI92-225EA 584 EUR InStock
PRSI92-225
Human recombinant 4-1BB Ligand (from E. coli)
Protéines et Peptides
tumour necrosis factor ligand superfamily member 9, also known as 4-1BBL, is a member of the the tumour necrosis factor family. It is a 254 amino acids cytokine that is expressed in brain, placenta, lung, skeletal muscle and kidney. TNFSF9 has been shown to reactivate anergic T lymphocytes in addition to promoting T lymphocyte proliferation. This cytokine has also been shown to be required for the optimal CD8 responses in CD8 T cells, and is thought to be involved in T cell-tumour cell interaction.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité