Human recombinant Cardiotrophin 1 (from E. coli)
Fournisseur: ProSci Inc.
92-226
PRSI92-226EA 789 EUR InStock
PRSI92-226
Human recombinant Cardiotrophin 1 (from E. coli)
Protéines et Peptides
Cardiotrophin-1, also known as CT-1 and CTF1, is a member of the IL-6 superfamily. It is a sreted cytokine that is expressed in heart, skeletal muscle, prostate and ovary, and to lower levels in lung, kidney, pancreas, thymus, testis and small intestine. The protein exerts its cellular effects by interacting with the glycoprotein 130 (gp130)/leukemia inhibitory factor receptor beta (LIFR) heterodimer. In addition, CT-1 activates phosphatidylinositol 3-kinase (PI-3 kinase) in cardiac myocytes and enhances transcription factor NF-κB DNA -binding activities.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité