Mouse recombinant CCL9 (from E. coli)
Fournisseur: ProSci Inc.
92-227
PRSI92-227EA 584 EUR InStock
PRSI92-227
Mouse recombinant CCL9 (from E. coli)
Protéines et Peptides
Mouse CCL9 is a small cytokine belonging to the CC chemokine family. It is expressed mainly in the liver, lung, and the thymus, although some expression has been detected in a wide variety of tissues except brain. CCL9 can activate osteoclasts through its receptor CCR1 (the most abundant chemokine receptor found on osteoclasts) suggesting an important role for CCL9 in bone resorption.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité