Human Recombinant Histatin-3 (from E. coli)
Fournisseur: ProSci Inc.
92-229
PRSI92-229EA 884 EUR InStock
PRSI92-229
Human Recombinant Histatin-3 (from E. coli)
Protéines et Peptides
HTN3 belongs to the histatin/statherin family. Histatins are salivary proteins that are considered to be major precursors of the protective proteinaceous structure on tooth surfaces (enamel pellicle). In addition, histatins exhibit antibacterial and antifungal activities. Post-translational proteolytic processing results in many histatins: e.g., histatins 4-6 are derived from histatin 3 by proteolysis. Histatins 1 and 3 are primary products of HIS1and HIS2 alleles, respectively. Histatins are believed to have important non-immunological, anti-microbial function in the oral cavity.
Fusion-Tag: N-GST tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité