Human recombinant Calcitonin gene-related peptide 2 (from E. coli)
Fournisseur: ProSci Inc.
92-230
PRSI92-230EA 409 EUR InStock
PRSI92-230
Human recombinant Calcitonin gene-related peptide 2 (from E. coli)
Protéines et Peptides
CALCB is a member of the calcitonin family. CALCB is produced in both peripheral and central neurons. It is a potent peptide vasodilator and can function in the transmission of pain. In the spinal cord, the function and expression of CGRP may differ depending on the location of synthesis. CALCB is derived mainly from the cell bodies of motor neurons when synthesised in the ventral horn of the spinal cord and may contribute to the regeneration of nervous tissue after injury.
Fusion-Tag: N-GST tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité