Human recombinant UBE2D3 (from E. coli)
Fournisseur: ProSci Inc.
92-231
PRSI92-231EA 306 EUR InStock
PRSI92-231
Human recombinant UBE2D3 (from E. coli)
Protéines et Peptides
UBE2D3 is an enzyme that belongs to the ubiquitin-conjugating enzyme family. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This enzyme functions in the ubiquitination of the tumour-suppressor protein p53, which is induced by an E3 ubiquitin-protein ligase.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité