Human recombinant ATPase SWSAP1 (from E. coli)
Fournisseur: ProSci Inc.
92-241
PRSI92-241EA 852 EUR InStock
PRSI92-241
Human recombinant ATPase SWSAP1 (from E. coli)
Enzymes
SWSAP1 is a nucleus ATPase protein, interacts with ZSWIM7 and forms a functional complex. The complexs involved in homologous recombination repair and stabilises each other. SWS1AP1 also interacts with RAD51, RAD51B, RAD51C, RAD51D and XRCC3. It involves in homologous recombination repair. ATPase is preferentially stimulated by single-stranded DNA and is involved in homologous recombination repair (HRR). SWSAP1 has a DNA-binding activity which is independent of its ATPase activity.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité