Human recombinant PSD95 (from E. coli)
Fournisseur: ProSci Inc.
92-242
PRSI92-242EA 654 EUR InStock
PRSI92-242
Human recombinant PSD95 (from E. coli)
Protéines et Peptides
Disks large homolog 4(DLG4) is a cell membrane protein and it is a member of the membrane-associated guanylate kinase (MAGUK) family. The protein contains 1 guanylate kinase-like domain,3 PDZ (DHR) domains and 1 SH3 domain. With PSD-93 it is recruited into the same NMDA receptor and potassium channel clusters. These two MAGUK proteins may interact at postsynaptic sites to form a multimeric scaffold for the clustering of receptors, ion channels, and associated signaling proteins. DLG4 is the best studied member of the MAGUK-family of PDZ domain-containing proteins. Like all MAGUK-family proteins, its basic structure includes three PDZ domains, an SH3 domain, and a guanylate kinase-like domain (GK) connected by disordered linker regions. It is almost exclusively located in the post synaptic density of neurons, and is involved in anchoring synaptic proteins. Its direct and indirect binding partners include neuroligin, NMDA receptors, AMPA receptors, and potassium channels.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité