Mouse recombinant EGF (from E. coli)
Fournisseur: ProSci Inc.
92-245
PRSI92-245EA 306 EUR InStock
PRSI92-245
Mouse recombinant EGF (from E. coli)
Protéines et Peptides
EGF is a single-pass type I membrane protein,containing 8 LDL-receptor class B repeats and 9 EGF-like domains. EGF results in cellular proliferation, differentiation, and survival.EGF is a low-molecular-weight polypeptide first purified from the mouse submandibular gland, but since then found in many human tissues including submandibular gland, parotid gland. Salivary EGF, which seems also regulated by dietary inorganic iodine, also plays an important physiological role in the maintenance of oro-esophageal and gastric tissue integrity. The biological effects of salivary EGF include healing of oral and gastroesophageal ulcers, inhibition of gastric acid secretion, stimulation of DNA synthesis as well as mucosal protection from intraluminal injurious factors such as gastric acid, bile acids, pepsin, and trypsin and to physical, chemical and bacterial agents.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité