Human recombinant IL18 (from E. coli)
Fournisseur: ProSci Inc.
92-246
PRSI92-246EA 654 EUR InStock
PRSI92-246
Human recombinant IL18 (from E. coli)
Protéines et Peptides
Interleukin-18 is a secreted protein and it belongs to the IL-1 family. IL-18 is a proinflammatory cytokine and produced by macrophages and other cells. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL-4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein (IL18BP) can specifically interact with this cytokine, and thus negatively regulate its biological activity. After stimulation with IL-18, natural killer (NK) cells and certain T cells release another important cytokine called interferon- gamma (IFN- gamma ) or type II interferon that plays an important role in activating the macrophages or other cells.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité