Human recombinant TMX2 (from E. coli)
Fournisseur: ProSci Inc.
92-247
PRSI92-247EA 852 EUR InStock
PRSI92-247
Human recombinant TMX2 (from E. coli)
Protéines et Peptides
TMX2 is a single-pass type I membrane protein and contains 1 thioredoxin domain. Thioredoxin plays an important role in various cellular processes through redox regulation. The molecular cloning and characterisation of one member of the thioredoxin superfamily, designated as TMX2.The TMX2 cDNA consists of 1644 nucleotides and contains an open reading frame encoding a protein of 372 amino acids with a predicted molecular mass of 42.5 kDa and an isoelectric point of 8.94. The TMX2 protein may possess an N-terminal signal peptide, a potential transmembrane domain, an Myb DNA-binding domain repeat signature, a thioredoxin consensus pattern, an endoplasmic reticulum (ER) membrane retention signal (KKXX-like motif), and a dileucine motif in the tail.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité