Human Recombinant EIF4E (from E. coli)
Fournisseur: ProSci Inc.
92-253
PRSI92-253EA 852 EUR InStock
PRSI92-253
Human Recombinant EIF4E (from E. coli)
Protéines et Peptides
Eukaryotic translation initiation factor 4E is a 217 amino acids protein that belongs to the eukaryotic initiation factor 4E family. eIF4F is a multi-subunit complex, the composition of which varies with external and internal environmental conditions. It is composed of at least EIF4A, EIF4E and EIF4G1/EIF4G3. EIF4E is also known to interact with other partners.It can recognize and bind the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité