Human recombinant TRIM5 (from E. coli)
Fournisseur: ProSci Inc.
92-254
PRSI92-254EA 884 EUR InStock
PRSI92-254
Human recombinant TRIM5 (from E. coli)
Protéines et Peptides
Tripartite motif-containing Motif 5 is a protein that in humans is encoded by the TRIM5 gene.It is a 493 amino acids protein that belongs to the TRIM/RBCC family.It contains 1 B box-type zinc finger, 1 B30.2/SPRY domain and 1 RING-type zinc finger. TRIM5 present in the cytoplasm recognises motifs within the capsid proteins and interferes with the uncoating process, therefore preventing successful reverse transcription and transport to the nucleus of the viral genome. The exact mechanism of action has not been shown conclusively, but capsid protein from restricted viruses is removed by proteasome-dependent degradation
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité