Human recombinant Intelectin-1 (from E. coli)
Fournisseur: ProSci Inc.
92-256
PRSI92-256EA 654 EUR InStock
PRSI92-256
Human recombinant Intelectin-1 (from E. coli)
Protéines et Peptides
Intelectin-1(ITLN1) is a secreted protein and contains 1 fibrinogen C-terminal domain. Intelectin-1 is a 40 kDa Ca-dependent galactofuranose-binding lectin that is not a C-type lectin. It is expressed on multiple cell types and appears to participate in insulin signaling and microbe recognition. The protein has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. It increases AKT phosphorylation in the absence and presence of insulin and it may play a role in the defense systemagainst microorganisms. It also may specifically recognize carbohydrate chains of pathogens and bacterial components containing galactofuranosy l residues, in a calcium-dependent manner.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité