Human Recombinant Repulsive guidance molecule A (from E. coli)
Fournisseur: ProSci Inc.
92-257
PRSI92-257EA 588 EUR InStock
PRSI92-257
Human Recombinant Repulsive guidance molecule A (from E. coli)
Protéines et Peptides
Repulsive guidance molecule A(RGMA) is a cell membrane protein and belongs to the repulsive guidance molecule (RGM) family. It interacts with NEO1, BMP2 and BMP4. RGMA is a glycosylphosphatidylinositol-anchored glycoprotein that functions as an axon guidance protein in the developing and adult central nervous system. It helps guide Retinal Ganglion Cell (RGC) axons to the tectum in the midbrain. RGMa has been implicated to play an important role in the developing brain and in the scar tissue that forms after a brain injury. This protein may also function as a tumor suppressor in some cancers.
  • Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays.
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité