Human Recombinant Dipeptidyl-peptidase 3 (from E. coli)
Fournisseur: ProSci Inc.
92-263
PRSI92-263EA 789 EUR InStock
PRSI92-263
Human Recombinant Dipeptidyl-peptidase 3 (from E. coli)
Enzymes
Dipeptidyl peptidase 3(DPP3), is a member of the S9B family in clan SC of the serine proteases. DPP3 has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH). It releases an N-terminal dipeptide from a peptide comprising four or more residues, with broad specificity and also acts on dipeptidyl 2-naphthylamides.Increased activity of this protein has a relationship with endometrial and ovarian cancers.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité