Human recombinant Ferritin light chain (from E. coli)
Fournisseur: ProSci Inc.
92-265
PRSI92-265EA 343 EUR InStock
PRSI92-265
Human recombinant Ferritin light chain (from E. coli)
Protéines et Peptides
Ferritin is a large, iron-storage heteropolymeric protein,which is expressed in most kinds of cells and co-assemble in different proportion in a tissue-specific manner. Ferritin has oligomer of 24 subunits and two types of subunits including light chain(FTL) and heavy chain. Ferritin can remove Fe (Ⅱ) from solution in the presence of oxygen and is very important for iron homeostasis. Iron is absorbed in the ferrous form and deposited as ferric hydroxides after oxidation. Iron is first oxidised to the ferric state for storage as ferric oxyhdroxide whithin the protein shell of ferritin. Thus, ferritin removes excess iron from the cell sap where it could otherwise participate in peroxidation mechanisms. Ferritin also plays a role in delivery of iron to cells and mediates iron uptake in capsule cells of the developing kidney.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité