Human recombinant Krueppel-like factor 6 (from E. coli)
Fournisseur: ProSci Inc.
92-270
PRSI92-270EA 852 EUR InStock
PRSI92-270
Human recombinant Krueppel-like factor 6 (from E. coli)
Protéines et Peptides
Krueppel-Like Factor 6 (KLF6) belongs to the krueppel C2H2-type zinc-finger protein family. KLF6 contains three C2H2-type zinc fingers and localises in the nucleus. KLF6 expression is highest in the placenta followed by spleen, thymus, prostate, testis, small intestinem and colon. However, it is weakly expressed in the pancreas, lung, liver, heart, and skeletal muscle. KLF6 functions as a transcriptional activator and could play a role in B-cell growth and development. Defects in KLF6 will result in gastric cancer and prostate cancer.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité