Human recombinant Annexin A3 (from E. coli)
Fournisseur: ProSci Inc.
92-273
PRSI92-273EA 852 EUR InStock
PRSI92-273
Human recombinant Annexin A3 (from E. coli)
Protéines et Peptides
Annexin A3(ANXA3)contains 4 annexin repeats and belongs to the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. It is abnormally expressed in fetuses of both IVF and ICSI, which may contribute to the increase risk of birth defects in these ART. ANXA3 is Inhibitor of phospholipase A2 and cleaves the cyclic bond of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. it also also play a role in anti-coagulant properties.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité