Human recombinant Peroxiredoxin 1 (from E. coli)
Fournisseur: ProSci Inc.
92-274
PRSI92-274EA 365 EUR InStock
PRSI92-274
Human recombinant Peroxiredoxin 1 (from E. coli)
Protéines et Peptides
Peroxiredoxin-1(PRDX1) contains 1 thioredoxin domain and belongs to the AhpC/TSA family. PRDX1 constitutively expressed in most human cells and it is induced to higher levels upon serum stimulation in untransformed and transformed cells. PRDX1 is involved in redox regulation of the cell. It reduces peroxides with reducing equivalents provided through the thioredoxin system but not from glutaredoxin and play an important role in eliminating peroxides generated during metabolism. PRDX1 might participate in the signaling cascades of growth factors and tumor necrosis factor-alpha by regulating the intracellular concentrations of H2O2. It reduces an intramolecular disulfide bond in GDPD5 that gates the ability to GDPD5 to drive postmitotic motor neuron differentiation. It may contribute to the antiviral activity of CD8(+) T-cells and have a proliferative effect in cancer development or progression.
Fusion-Tag: N, C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité