Human recombinant Activating Transcription Factor 1 (from E. coli)
Fournisseur: ProSci Inc.
92-277
PRSI92-277EA 852 EUR InStock
PRSI92-277
Human recombinant Activating Transcription Factor 1 (from E. coli)
Protéines et Peptides
Cyclic AMP-dependent transcription factor ATF-1(ATF1) which contains 1 bZIP (basic-leucine zipper) domain and 1 KID (kinase-inducible) domain, belongs to the bZIP family. It influences cellular physiologic processes by regulating the expression of downstream target genes, which are related to growth, survival, and other cellular activities. ATF1 binds the cAMP response element (CRE) (consensus: 5'-GTGACGT[AC][AG]-3'), a sequence present in many viral and cellular promoters. It also binds to the Tax-responsive element (TRE) of HTLV-I. ATF1 mediates PKA-induced stimulation of CRE-reporter genes, represses the expression of FTH1 and other antioxidant detoxification genes, triggers cell proliferation and transformation. ATF1 is phosphorylated at serine 63 in its kinase-inducible domain by serine/threonine kinases, cAMP-dependent protein kinase A, calmodulin-dependent protein kinase I/II, mitogen- and stress-activated protein kinase and CDK3. Its phosphorylation enhances its transactivation and transcriptional activities, and enhances cell transformation.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité