Human Recombinant Heat Shock beta 2 (from E. coli)
Fournisseur: ProSci Inc.
92-279
PRSI92-279EA 306 EUR InStock
PRSI92-279
Human Recombinant Heat Shock beta 2 (from E. coli)
Protéines et Peptides
Heat shock protein beta-2(HSPB2) is a protein that in humans is encoded by the HSPB2 gene. HSPB2 belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. It is expressed preferentially in the heart and skeletal muscle. HSPB2 has been shown to interact with TRAF6, HSPB8, Myotonic dystrophy protein kinase and CRYAB. HSPB2 regulates Myotonic Dystrophy Protein Kinase, which plays an important role in maintenance of muscle structure and function.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité