Human recombinant Methenyl-THF synthetase (from E. coli)
Fournisseur: ProSci Inc.
92-280
PRSI92-280EA 789 EUR InStock
PRSI92-280
Human recombinant Methenyl-THF synthetase (from E. coli)
Enzymes
5-formyltetrahydrofolate cyclo-ligase (MTHFS) belongs to the 5-formyltetrahydrofolate cyclo-ligase family. It is an enzyme that catalyses the conversion of 5-formyltetrahydrofolate to 5,10-methenyltetrahydrofolate, contributes to tetrahydrofolate metabolism. MTHFS helps regulate carbon flow through the folate-dependent one-carbon metabolic network that supplies carbon for the biosynthesis of purines, thymidine and amino acids. An increased activity of the encoded protein can result in an increased folate turnover rate and folate depletion.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité