Mouse recombinant S100 calcium binding A8 (from E. coli)
Fournisseur: ProSci Inc.
92-283
PRSI92-283EA 654 EUR InStock
PRSI92-283
Mouse recombinant S100 calcium binding A8 (from E. coli)
Protéines et Peptides
Protein S100-A8(Mrp8) contains 2 EF-hand domains and belongs to the S-100 family. Mrp8 binds two calcium ions per molecule with an affinity similar to that of the S-100 proteins. S100 proteins are localised in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. It may function in the inhibition of casein kinase and as a cytokine. Altered expression of this protein is associated with the disease cystic fibrosis.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité