Human Recombinant 40S Ribosomal S7 (from E. coli)
Fournisseur: ProSci Inc.
92-289
PRSI92-289EA 982 EUR InStock
PRSI92-289
Human Recombinant 40S Ribosomal S7 (from E. coli)
Protéines et Peptides
40S ribosomal protein S7(RPS7) belongs to the S7E family of ribosomal proteins. It is phosphorylated by NEK6 during post-translational modification. RPS7 is located in the cytoplasm, binds IPO9 with high affinity. it also can interacts with NEK6. As is required for rRNA maturation and typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. The unnormal expression of RPS7 may cause Diamond-Blackfan anemia 8.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité