Human Recombinant Heat Shock beta 8 (from E. coli)
Fournisseur: ProSci Inc.
92-290
PRSI92-290EA 884 EUR InStock
PRSI92-290
Human Recombinant Heat Shock beta 8 (from E. coli)
Protéines et Peptides
Heat shock protein beta-8 (HSPB8) belongs to the small heat shock protein (HSP20) family. This protein can be inducted by 17-beta-estradiol, and is predominantly expressed in skeletal muscle and heat, mainly located in the cytoplasm and nucleus. HSPB8 usually exists in monomer, it can interact with HSPB1 and DNAJB6. HSPB8 displays temperature-dependent chaperone activity,appears to be involved in regulation of cell proliferation, apoptosis, and carcinogenesis, and mutations in this gene have been associated with different neuromuscular diseases, including Charcot-Marie-Tooth disease.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité