Human recombinant PDGF-AA (from E. coli)
Fournisseur: ProSci Inc.
92-294
PRSI92-294EA 852 EUR InStock
PRSI92-294
Human recombinant PDGF-AA (from E. coli)
Protéines et Peptides
Platelet-derived growth factor subunit A (PDGFA), belongs to the PDGF/VEGF growth factor family. PDGFA is a secreted protein, stored in platelet alpha-granules and released by platelets upon wounding. PDGFA is potent mitogens for a variety of cell types including smooth muscle cells, connective tissue cells, bone and cartilage cells, and some blood cells. It plays an essential role in the regulation of embryonic development, cell proliferation, cell migration, survival and chemotaxis. PDGFA is required for normal lung alveolar septum formation during embryogenesis, normal development of the gastrointestinal tract, normal development of Leydig cells and spermatogenesis, normal oligodendrocyte development and normal myelination in the spinal cord and cerebellum. It plays an important role in wound healing; Signaling is modulated by the formation of heterodimers with PDGFB.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité