Human recombinant Cyclin-H (from E. coli)
Fournisseur: ProSci Inc.
92-306
PRSI92-306EA 654 EUR InStock
PRSI92-306
Human recombinant Cyclin-H (from E. coli)
Protéines et Peptides
Human CCNH, also known as Cyclin-H,MO15-associated protein,p34 and p37, is a protein which belongs to the highly conserved cyclin family. Cyclin family members are characterised by a dramatic periodicity in protein abundance through the cell cycle. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin forms a complex with CDK7 kinase and ring finger protein MAT1.CCNH regulates CDK7 which is the catalytic subunit of the CDK-activating kinase (CAK) enzymatic complex. CAK activates the cyclin-associated kinases CDK1, CDK2, CDK4 and CDK6 by threonine phosphorylation. CAK complexed to the core-TFIIH basal transcription factor activates RNA polymerase II by serine phosphorylation of the repetitive C-terminal domain (CTD) of its large subunit (POLR2A), allowing its escape from the promoter and elongation of the transcripts. CCNH is also involved in cell cycle control and in RNA transcription by RNA polymerase II. Its expression and activity are constant throughout the cell cycle.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité