Human recombinant Acylphosphate phosphohydrolase 1 (from E. coli)
Fournisseur: ProSci Inc.
92-308
PRSI92-308EA 365 EUR InStock
PRSI92-308
Human recombinant Acylphosphate phosphohydrolase 1 (from E. coli)
Enzymes
ACYP1, also known as Acylphosphatase-1, Acylphosphatase, erythrocyte isozyme, Acylphosphatase, organ-common type isozyme, Acylphosphate phosphohydrolase 1 and ACYPE, is a small cytosolic enzyme which catalyses the hydrolysis of the carboxyl-phosphate bond of acylphosphates. ACYP1 is a protein which belongs to the acylphosphatase family and contains 1 fibrinogen C-terminal domain. Two isoenzymes have been isolated, called muscle acylphosphatase and erythrocyte acylphosphatase, on the basis of their tissue localisation. This gene encodes the erythrocyte acylphosphatase isoenzyme. Alternatively spliced transcript variants that encode different proteins were identified through data analysis. Recombinant human ACYP1 protein was expressed in E. coli fused with HIS-tag at N-terminus.
Fusion-Tag: N-6 His tag.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité