Human recombinant IL7RA (from cells)
Fournisseur: ProSci Inc.
92-313
PRSI92-313EA 588 EUR InStock
PRSI92-313
Human recombinant IL7RA (from cells)
Protéines et Peptides
Interleukin 7 Receptor alpha (IL-7R alpha), also known as CD127, is a 75 kDa hematopoietin receptor superfamily member that plays an important role in lymphocyte differentiation, proliferation, and survival. IL-7R alpha is majorly expressed on T cells and their precursors, and early in B cell development as well, prior to the appearance of surface IgM. Dynamic regulation of IL-7R alpha is important for the generation of appropriate immune responses.
Fusion-Tag: C-Fc-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité