Human recombinant EPDR1 (from cells)
Fournisseur: ProSci Inc.
92-314
PRSI92-314EA 409 EUR InStock
PRSI92-314
Human recombinant EPDR1 (from cells)
Protéines et Peptides
EPDR1 is a member of the ependymin family. EPDR1 is a type II transmembrane protein that is similar to two families of cell adhesion molecules, the protocadherins and ependymins. It may play a role in calcium-dependent cell adhesion. EPDR1 is glycosylated, and the orthologous mouse protein is localised to the lysosome. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 8.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité