Human recombinant Methyl-CpG-binding 2 (from cells)
Fournisseur: ProSci Inc.
92-318
PRSI92-318EA 761 EUR InStock
PRSI92-318
Human recombinant Methyl-CpG-binding 2 (from cells)
Protéines et Peptides
The MeCP2 helps regulate gene activity (expression) by modifying chromatin, the complex of DNA and protein that packages DNA into chromosomes. The MeCP2 protein is present in cells throughout the body, although it is particularly abundant in brain cells.In the brain, the MeCP2 protein likely plays a role in maintaining connections (synapses) between neurons, where cell-to-cell communication occurs. The alternative splicing of proteins is critical for normal communication between neurons and may also be necessary for the function of other types of brain cells.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité