Human recombinant Pancreatic Polypeptide (from cells)
Fournisseur: ProSci Inc.
92-320
PRSI92-320EA 761 EUR InStock
PRSI92-320
Human recombinant Pancreatic Polypeptide (from cells)
Protéines et Peptides
PPY belongs to the NPY family and is Synthesised as a 95 aa polypeptide precursor in the pancreatic islets of Langerhans. It is cleaved into two peptide products; the active hormone of 36 aa and an icosapeptide of unknown function. The hormone acts as a regulator of pancreatic and gastrointestinal functions and may be important in the regulation of food intake. Plasma level of this hormone has been shown to be reduced in conditions associated with increased food intake and elevated in anorexia nervosa.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité