Mouse recombinant Testican 3
Fournisseur: ProSci Inc.
92-323
PRSI92-323EA 525 EUR InStock
PRSI92-323
Mouse recombinant Testican 3
Protéines et Peptides
SPOCK3,also called Testican3,is a secreted protein and make up a family of extracellular heparan/chondroitin sulphate proteoglycans. It contains 1 Kazal-like domain and 1 thyroglobulin type-1 domain and mainly expressed in brain. SPOCK3 contain inhibitory regions in several domains targeted to different classes of protease, and in some cases may act as protease inhibitors. In addition to their presence in testis, testicans are enriched in brain and have been shown to regulate neuronal attachment and outgrowth.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité