Human recombinant magnesium-dependent phosphatase 1 (from cells)
Fournisseur: ProSci Inc.
92-327
PRSI92-327EA 789 EUR InStock
PRSI92-327
Human recombinant magnesium-dependent phosphatase 1 (from cells)
Enzymes
MDP1, which is short for nesium-dependent phosphatase 1, is a 176 aa. protein. It has 3 forms by different alternative splicing. This protein belongs to the HAD-like hydrolase superfamily, and usually takes its function with Magnesium. It can be inhibited by vanadate and zinc, and slightly by calcium. MDP1 is a Magnesium-dependent phosphatase which may act as a tyrosine phosphatase.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité