Human recombinant VSTM2A (from cells)
Fournisseur: ProSci Inc.
92-330
PRSI92-330EA 852 EUR InStock
PRSI92-330
Human recombinant VSTM2A (from cells)
Protéines et Peptides
VSTM2A, also called VSTM2, is short for V-set and transmembrane domain-containing protein 2A. It is a 236 aa. protein which can be departed into two parts. The first 24 aa. is a signal peptide, and the last is a V-set and transmembrane domain-containing protein 2A. There is a natural variant location which is E84K.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité