Human recombinant ITI heavy chain H3 (from cells)
Fournisseur: ProSci Inc.
92-332
PRSI92-332EA 852 EUR InStock
PRSI92-332
Human recombinant ITI heavy chain H3 (from cells)
Protéines et Peptides
ITIH3, which is short for Inter-alpha-trypsin inhibitor heavy chain H3, is a 890 aa. protein. It is secreted expression, and belongs to the ITIH family. I-alpha-I plasma protease inhibitors are assembled from one or two heavy chains (H1, H2 or H3) and one light chain, bikunin. Inter-alpha-inhibitor (I-alpha-I) is composed of H1, H2 and bikunin, inter-alpha-like inhibitor (I-alpha-LI) of H2 and bikunin, and pre-alpha-inhibitor (P-alpha-I) of H3 and bikunin. ITTH3 may act as a carrier of hyaluronan in serum or as a binding protein between hyaluronan and other matrix protein, including those on cell surfaces in tissues to regulate the localisation, synthesis and degradation of hyaluronan which are essential to cells undergoing biological processes.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité