Human recombinant TXNDC15 (from cells)
Fournisseur: ProSci Inc.
92-333
PRSI92-333EA 761 EUR InStock
PRSI92-333
Human recombinant TXNDC15 (from cells)
Protéines et Peptides
TXNDC15, which is short for oredoxin domain-containing protein 15, is encoded by FAM172A gene. It is also known as C5orf14. TXNDC15 is a 360 aa. protein, and has 2 isoforms produced by alternative splicing. There is a natural variant location which is S248P and it has a 18 aa. signal peptide. TXNDC15 contains 1 thioredoxin domain. This protein is a Single-pass type I membrane protein and locates at membrane.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité